missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Mevalonolactone, 97%, Thermo Scientific Chemicals
£84.70 - £341.00
Chemical Identifiers
CAS | 674-26-0 |
---|---|
Molecular Formula | C6H10O3 |
Molecular Weight (g/mol) | 130.14 |
MDL Number | MFCD00006648 |
InChI Key | JYVXNLLUYHCIIH-UHFFFAOYNA-N |
Synonym | mevalonolactone, dl-mevalonic acid lactone, dl-mevalonolactone, 4-hydroxy-4-methyltetrahydro-2h-pyran-2-one, mevalonic acid lactone, mevalonic lactone, dl-mevalolactone, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl-, .+/-., 1-tetrahydro-4-hydroxy-4-methyl-2h-pyran-2-one |
PubChem CID | 10428 |
IUPAC Name | 4-hydroxy-4-methyloxan-2-one |
SMILES | CC1(O)CCOC(=O)C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10422790
|
Thermo Scientific Acros
428300010 |
1 g | Glass Bottle |
£84.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10119699
|
Thermo Scientific Acros
428300050 |
5 g | Glass Bottle |
£341.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
674-26-0 | |
130.14 | |
JYVXNLLUYHCIIH-UHFFFAOYNA-N | |
10428 | |
CC1(O)CCOC(=O)C1 |
C6H10O3 | |
MFCD00006648 | |
mevalonolactone, dl-mevalonic acid lactone, dl-mevalonolactone, 4-hydroxy-4-methyltetrahydro-2h-pyran-2-one, mevalonic acid lactone, mevalonic lactone, dl-mevalolactone, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl-, .+/-., 1-tetrahydro-4-hydroxy-4-methyl-2h-pyran-2-one | |
4-hydroxy-4-methyloxan-2-one |
Specifications
674-26-0 | |
100.0 | |
145.0°C to 150.0°C (5.0 mmHg) | |
96% min. (GC) | |
MFCD00006648 | |
Solubility in water: soluble | |
CC1(O)CCOC(=O)C1 | |
130.14 | |
130.14 | |
(±)-Mevalonolactone |
96.0 | |
28.0°C | |
>110°C | |
C6H10O3 | |
mevalonolactone, dl-mevalonic acid lactone, dl-mevalonolactone, 4-hydroxy-4-methyltetrahydro-2h-pyran-2-one, mevalonic acid lactone, mevalonic lactone, dl-mevalolactone, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl-, .+/-., 1-tetrahydro-4-hydroxy-4-methyl-2h-pyran-2-one | |
JYVXNLLUYHCIIH-UHFFFAOYNA-N | |
4-hydroxy-4-methyloxan-2-one | |
10428 | |
97% |
Safety and Handling
missing translation for 'einecsNumber' : 211-615-
RUO – Research Use Only