Learn More
Gilson™ MF1 Pump Head for Gilson™ Peristaltic Pump
Brand:  Gilson™ F110705
Code : A0
Additional Details : Weight : 0.01000kg
Product Code. 12396112
  • £605.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
Add channels to your MINIPULS™ 3 or Evolution and allow even greater flexibility in your laboratory.
MINIPULS 3 Pump Heads
Choose from these MINIPULS 3 pump heads:
  • R1—Single channel pump head
  • R4—Four channel pump head
  • R8—Eight channel pump head
  • R2—Two channel pump head
  • R2HF—Two channel high flow pump head
  • R4HF—Four channel high flow pump head
MINIPULS Evolution Pump Heads
Choose from these MINIPULS Evolution pump heads:
  • MF1—Pump head with one cartridge
  • MF4—Pump head with four cartridges
  • HF2—Pump head with two high flow cartridges
Specifications
Specifications
0.2μL/min. to 70 mL/min.