missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR2 (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577419
Additional Details : Weight : 0.01000kg
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Reconstitute using 50 uL deionized water.
Glutamate is the main excitatory neurotransmitter in the brain. For many years it had been considered to act only on the ligand-gated receptor channels-termed NMDA, AMPA and kainite receptors that are involved in the fast excitatory synaptic transmission. Recently, glutamate has been shown to regulate enzymes producing second messengers via specific receptors coupled to G-proteins. These receptors are called metabotropic glutamate receptors. In expression systems, Group-I receptors stimulate phospholipase C as revealed by an increase in phosphoinositide turnover and calcium release from internal stores. Group-II and -III receptors are coupled to the inhibition of adenylyl cyclase. The Group-I receptors include mGluR1a and mGluR5. The Group-II receptors in include mGluR2 and mGluR3.Specifications
mGluR2 (extracellular) | |
Polyclonal | |
Unconjugated | |
GRM2 | |
GRM2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
108068, 24415, 2912 | |
-20°C | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
P31421, Q14416, Q14BI2 | |
Grm2 | |
Peptide SLSRGADGSRHIC, corresponding to amino acids 109-121 of rat mGluR2 | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |