missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Mibefradil dihydrochloride
CaV3.x blocker
£218.00 - £788.00
Specifications
Purity | >97% |
---|---|
Product Type | Mibefradil dihydrochloride |
Molecular Weight (g/mol) | 568.56 |
Inhibitors | Mibefradil dihydrochloride |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785776
|
Tocris Bioscience™
2198/10 |
10 mg |
£218.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18093982
|
Tocris Bioscience™
2198/50 |
50 mg |
£788.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ca2+ channel blocker with moderate selectivity for T-type Ca2+ channels displaying IC50 values of 2.7 μM and 18.6 μM for T-type and L-type channels respectively. Antihypertensive agent.Specifications
>97% | |
Mibefradil dihydrochloride | |
568.56 | |
Mibefradil dihydrochloride |
For Research Use Only