missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miconazole (Free Base), Fisher BioReagents
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Amber Glass
Description
Miconazole is used as an antifungal agent.
Specifications
Specifications
Type | Miconazole |
Physical Form | Solid |
Melting Point | 135°C |
Ignition Residue | < 0.2% |
Assay Percent Range | 98% |
Loss on Drying | < 0.5% |
Merck Index | 15, 6257 |
Packaging | Amber Glass |
IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole |
Formula Weight | 416.13 |
Show More |
Safety and Handling
Product Identifier
- Miconazole
- Danger
- Acute toxicity Category 4
- Skin sensitiser Category 1
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'