missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU VitraPOR™ Micro-Filter Funnel
Micro-Filter Funnel for very small volumes.
Brand: ROBU VitraPOR™ 27203
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
2 mL | |
Borosilicate Glass (3.3) | |
3 | |
5/Pk. |