missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Micro-Filter Funnel

Micro-Filter Funnel for very small volumes.

Brand:  ROBU VitraPOR™ 27080

 View more versions of this product

Product Code. 16873066

  • £103.00 / Pack of 5
Estimated Shipment: 11-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

0.8 mL
Borosilicate Glass (3.3)
0
5/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers