missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Micro-Ninety™ Cleaning Solution

Use this concentrated cleaning solution for critical applications. Cole-Parmer™ Micro-Ninety™ Cleaning Solution cleans glass, most metals, plastic, rubber, fabric and is safe for all ultrasonic cleaners.

Brand:  Cole-Parmer™ 18100-05

Code : NEW

Additional Details : Weight : 1.08000kg


Product Code. 12061699

  • £25.28 / Each
In Stock
to see stock.
Add to basket

Description

Description

  • A mild, yet powerful, free-rinsing cleaner for use in critical cleaning and general purpose applications
  • Radioactive contaminants will not reduce cleaning capability
  • Removes most biological materials
  • Solution works at any concentration and rinses clean; most applications require only 1.25 to 2.5 fluid ounces per gallon of water
  • Solution is environmentally friendly
  • Contains no phosphates, CFCs, ODCs or borates
  • It safely replaces chromic acid to remove protein from glassware
  • Note: Do not use on zinc, magnesium or aluminum 
Specifications

Specifications

Liquid
Yellow
Bottle
9.5
100°C
1 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • micro-90 concentrared cleaning solution

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : EDTA Na4

SDS
Documents

Documents

Certificates
Special Offers

Special Offers