missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ Micro pH Electrodes
pH electrodes designed for small sample volumes or for measuring in small containers.
Brand: METTLER TOLEDO™ 51343160
314.93 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. (Connection cable is not included; sold separately.)
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Vials
- Microplates
- Test tubes
- Serum and Gastric juice
- Micro-biological samples
- Paint
- Gel
- Soap
- Shampoo
- Cosmetics
Electrode Specifications
- InLab Micro: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- InLab Micro Pro-ISM: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- InLab Ultra-Micro-ISM: ceramic junction; refillable with FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
- InLab Semi-Micro: ceramic junction; non-refillable - replace when electrolyte depleted
- InLab Semi-Micro-L: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- InLab Flex-Micro: porous PTFE junction; non-refillable - replace when electrolyte depleted
Accessories & Care
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
S7 | |
Glass | |
0°C to 80°C | |
3mol/L KCl | |
Requires Cable, S7 to BNC (30281915) | |
U Glass | |
<20 sec. | |
32°F to 176°F | |
60 mm | |
3 mm |
Ceramic | |
0 to 14 | |
Combination Electrode | |
Measure pH in the smallest vessels used in the laboratory | |
ARGENTHAL with Ag+ trap | |
InLab™ Micro | |
3mol/L KCl | |
2.36 in. | |
0.12 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction