Learn More
METTLER TOLEDO™ Micro pH Electrodes
pH electrodes designed for small sample volumes or for measuring in small containers.
Brand: METTLER TOLEDO™ 30244732
378.80 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.
Additional Details : Weight : 0.13500kg
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. (Connection cable is not included; sold separately.)
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Vials
- Microplates
- Test tubes
- Serum and Gastric juice
- Micro-biological samples
- Paint
- Gel
- Soap
- Shampoo
- Cosmetics
Electrode Specifications
- InLab Micro: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- InLab Micro Pro-ISM: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- InLab Ultra-Micro-ISM: ceramic junction; refillable with FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
- InLab Semi-Micro: ceramic junction; non-refillable - replace when electrolyte depleted
- InLab Semi-Micro-L: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- InLab Flex-Micro: porous PTFE junction; non-refillable - replace when electrolyte depleted
Accessories & Care
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
pH Electrode InLab Ultra-Micro-ISM | |
Ceramic | |
1 to 11 | |
Measuring small sample volumes in very small vessels such as vials, micro plates and cuvettes | |
ARGENTHAL™ with Ag+-trap | |
InLab™ Ultra-Micro-ISM | |
InLab storage solution | |
15 μL | |
40 mm |
Multi-Pin | |
Glass | |
Combination Electrode; Integrated Temperature Probe; Intelligent Sensor Management (ISM) Technology | |
Requires Cable, Multi-Pin to BNC/RCA (30281896) | |
LoT Glass | |
<20 sec. | |
0°C to 80°C | |
40 mm | |
3 mm |