missing translation for 'onlineSavingsMsg'
Learn More
Ansell MICRO-TOUCH Affinity Gloves Product Code.: 16306901
In Stock
to see stock.
Add to basket

Ansell MICRO-TOUCH Affinity Gloves

Product Code. 16306901 Shop All Ansell Products
Small, 6.5/7
Click to view available options
Size:
Large, 8.5/9
Medium, 7.5/8
Small, 6.5/7
X-Large, 9.5/10
This item is not returnable. View return policy

Product Code. 16306901

Brand: Ansell 553352

In Stock
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

MICRO-TOUCH Affinity Gloves are neoprene, powder-free examination gloves with excellent elasticity and barrier protection.

  • Robust Neoprene formulation provides comfort and excellent tear and puncture resistance
  • Textured fingers for secure grip
  • Protects from Type I latex allergy
  • Tested for use with chemotherapy drugs

Specifications

Accelerator Free Yes
Applications Laboratory
Color Green
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Polychloroprene
Powdered Powder Free
Product Type Surgical Gloves
Size Small, 6.5/7
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell MICRO-TOUCH Affinity Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.