missing translation for 'onlineSavingsMsg'
Learn More

3M™ Microbial Luminescence System Injector Cleaning Kit 3005 DPQCOG-CLNKIT

Injector cleaning kit includes 10 ATP Free Water and 2 Clean Solutions.

Brand:  3M™ 7000043164

Encompass

Product Code. 17293724

  • £121.04 / Each
Estimated Shipment: 10-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For daily use.
  • Prevents biofilm build-up.
  • Contains: 2 cleaning solutions (125 mL each), 10 ATP-free water (125 mL each) and 10 amber vials.
  • Storage: 2-8°C.
Specifications

Specifications

10 ATP Free Water, 2 Clean Solutions and 10 amber vials
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers