Learn More
3M™ Microbial Luminescence System Microwell Plate 3007
A Microwell Plate for use with the 3M™ Microbial Luminescence System (MLS).
Brand: 3M™ 7000042943
This item is not returnable.
View return policy
Description
- Microbial Luminescence System Microwell Plate 3007
- Used primarily for reagent checks, low-volume UHT testing
- Nonreusable microwell plate
Specifications
50/Cs. | |
3 M™ Microbial Luminescence System (MLS) |