Learn More
Neogen Microbial Luminescence System Microwell Strip 3008
Product Code. 17208530
Encompass
Click to view available options
Packaging Quantity:
320/Cs.
Unit Size:
Pack of 200
Product Code. 17208530
Brand: Neogen 7000030320
Microwell strip for use with the Neogen Microbial Luminescence System (MLS).
  • Plastic strips of 12 wells.
  • Snap to desired number of wells.
  • Use with Neogen MLS Microwell Strip Holder #3009.
  • Used primarily for reagent checks and low-volume UHT testing.
Specifications
Product Type Plastic Strips of 12 Wells
Packaging Quantity 320/Cs.
For Use With (Equipment) 3M™ MLS Microwell Strip Holder #3009.