Learn More
3M™ Microbial Luminescence System Ultra High Temperature Beverage Screen Kit BEV600
A beverage screen kit used with the 3M™ Microbial Luminescence System (MLS) for testing Ultra high temperature (UHT) and ESL beverages.
Brand: 3M™ 7100084525
Additional Details : CAS Number : 107-97-1 Weight : 0.00200kg
Description
- reagents for 600 tests.
- Eliminates second incubation period needed with traditional methods
Specifications
Microbiology Media | |
M™ Microbial Luminescence System (MLS) | |
600 |
Safety and Handling
- X600 MLS UHT Beverage Screen Kit - BEV600
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Cationic Agent, HEPES Acid, HEPES Salt