missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Microscope Slide Storage Box

Store up to 108 microscope slides. Azpack™ Microscope Slide Storage Box can protect up to 9 slides per tray.

Brand:  Azpack™ 12663845

Product Code. 12663845

  • £127.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

  • Holds up to 108 microscope slides 76 x 26mm
  • Cloth banded cardboard
  • Drop down front
  • Includes 12 loose trays
Specifications

Specifications

Slide Storage Box
Slides
Microscope slides
(12) Loose Tray, Drop Down Front, Lid
76 mm
Rectangular
26 mm
108
Storage
67 mm
76 mm
Cardboard
26 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.