missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labconco™ Microcentrifuge Tube Holder
Provides temperature stability to small samples that are susceptible to meltback. Labconco™ Microcentrifuge Tube Holder accommodates 1.7mL microcentrifuge tubes
Brand: Labconco™ 7439300
Code : 50
Additional Details : Weight : 2.27000kg
Description
- Anodized aluminum block with twelve bore holes that accommodate 1.7mL microcentrifuge tubes
- Holder may be placed on any flat freeze drying surface such as inside Clear Chambers or the built-in vacuum drying chamber of FreeZone Console Systems; or on FreeZone Stoppering or Bulk Tray Dryer Shelves, Product Shelves, or Heated Product Shelves
- Gives temperature stability to small samples that are susceptible to meltback
- Microcentrifuge tubes are not included
Specifications
Microcentrifuge Tube Holder | |
76.2 mm | |
25.4 mm | |
57.15 mm |
3.0 in. | |
1.0 in. | |
2.25 in. |