missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Life Sciences™ MicroCheck II Beverage Monitor

Easy to use, versatile, disposable filter funnels for microbial analysis.

Brand:  Cytiva Life Sciences™ 4761ME

Product Code. 17977262

  • £143.03 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Can convert to a Petri dish for culturing or easily remove the membrane for placing on separate agar dish.

  • Can convert to a Petri dish for culturing or easily remove the membrane for placing on separate agar dish.
  • Ready to use: simply remove from box and place directly on a filter manifold.
  • No need for rubber stoppers or adapters.
  • Volume graduations are marked clearly for easy reading from any point of view.
  • Volume graduations marked externally on the funnel to prevent potential hold-up of organisms.
  • External marks meet guidelines listed in Standard Methods for the Analysis of Water and Wastewater, current edition.
  • Filter Media: GN Metricel™ (mixed cellulose esters) and Metricel™ Black (modified polyethersulfone) membranes
  • Cylinder and Base: Polypropylene
  • Cover: Polystyrene
  • Plug: Polyethylene
  • Funnel Adapter: Polyethylene
  • Support Pad: Cellulose
  • Height: 7.6 cm (3.0 in.)
  • Diameter: 6.1 cm (2.4 in.)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL
Beverage monitor
50 Funnels
Filtering Funnel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cytiva Life Sciences™ MicroCheck II Beverage Monitor > Cytiva Life Sciences™ Beverage Monitor

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.