missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ MICROCHEM™ 2000 Oversleeves, Model 600
Click to view available options
Size: One Size
Description
- Proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards
- Optimized fit, improving wearer comfort and safety
- Low Linting
- Ideal for clean room cleaning and preparing, production line support and maintenance, and maintenance of plant and machines
Specifications
Size | Universal |
Product Type | Oversleeves |
Material | Non-woven Microporous Polyethylene Laminate |
Color | White |
Style | Elastic at Both Ends |
Seam Type | Bound |
Certifications/Compliance | CE Category I |
Length (English) | 20 in. |
Length (Metric) | 508 mm |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'