Learn More
Ansell™ Microchem™ 3000 Aprons with Sleeves
missing translation for 'orderingAttributeHoverText'
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Apron with Sleeves
- Rear velcro fastening
- Elasticated wrists
- Welded seams
- Multi-layer barrier fabric effective against numerous chemicals
- Highly visible bright yellow for improved worker safety
- Lightweight yet durable
- Anti-static according to EN 1149-5
Specifications
Specifications
Type | Protective Apron |
Size | 2X-Large |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
Closure Type | Hook and Loop (Rear) |
Cuff Style | Elastic |
No. of Pockets | None |
Certifications/Compliance | Type PB 3/4 EN 14605, Type PB(3)-B, PB(4)-B EN 14126, EN1149-1 |
Seam Type | Ultrasonically Welded |