missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Microchem™ 3000 Aprons with Sleeves
Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 40
Pack of 45
Description
- Lightweight and durable
Specifications
Specifications
| Size | 3X-Large |
| Material | Multi-layer Barrier Laminate Fabric |
| Color | Yellow |
| Closure Type | Buckles (Rear) |
| For Use With (Application) | Unexpected Leakages Spills or Other Releases, Loading and Unloading Trucks and Vehicles, Loading Trucks & Vehicles, Product Inspection, Cleaning of Plant and Machines |
| Weight (Metric) | 80 gsm |
| Style | Y30W214 |
| Seam Type | Welded |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction