missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Microchem™ 4000 Cape Hood with Visor

Attaches under arms with hook-and-loop closure straps. Ansell Edmont™ Microchem™ 4000 Cape Hood with Visor protects from organic and inorganic chemical and biological agents.

Brand:  Ansell Edmont™ GR40T-00516-00

980.91 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 15536464

  • £1435.00 / Pack of 20
Estimated Shipment: 10-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Constructed of multi-layer barrier laminate fabric
  • Hood with PVC visor and 76mm body panel
  • Welded and taped seams
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Protective Hood
Multi-layer Barrier Laminate Fabric, PVC
20/Pk.
Universal
Green
Ultrasonically welded
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell Edmont™ Microchem™ 4000 Cape Hood with Visor >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.