Learn More
Ansell™ Microchem™ 4000 PAPR Encapsulated Coveralls
MICROCHEM™ 4000 PAPR Encapsulated Coveralls provide complete protection from liquid and particulate hazards. Ventilated suits with filtered air and MICROGARD™/MICROCHEM™ technology provide head and body protection from hazardous substances.
£614.00
Specifications
Certifications/Compliance Type 3/4 EN14605, Type 5 EN ISO 13982-1, Types 3-B, 4-B EN14126, EN1073-2, EN1149-5
Color Green
Hood Type Fully Encapsulated
Material Multi-layer Barrier Laminate
Ankle Style Attached socks, Boot overflaps
View More Specs
Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15546464
View Documents
Ansell™
GR40T-00701-03
Medium This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15556464
View Documents
Ansell™
GR40T-00701-04
Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15566464
View Documents
Ansell™
GR40T-00701-05
X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15576464
View Documents
Ansell™
GR40T-00701-06
2X-Large This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Fully Encapsulated Coverall, for use in combination with Scott Proflow 2SC/EX PAPR fan unit, breathing hose & filters.
  • Double elasticated cuffs, attached socks with boot overflap.
  • Dual zip front entry
  • Emergency rip cords attached to left chest shoulder and left back shoulder as worn.
  • Permeation tested against over 180 chemicals, including chemical warfare agents
  • Textile-like inner provides wearer acceptance
  • Anti-static according to EN 1149-5
Specifications
Specifications
Type 3/4 EN14605, Type 5 EN ISO 13982-1, Types 3-B, 4-B EN14126, EN1073-2, EN1149-5
Fully Encapsulated
Attached socks, Boot overflaps
Double zipper front entry
Emergency rip cords, attached socks with boot overflap
Welded, Taped
Green
Multi-layer Barrier Laminate
Accommodates Scott Proflow 2SC/EX PAPR fan unit, breathing hose and filters
Double, Elastic
None
Includes Emergency rip cords, attached socks with boot overflap