Learn More
Thermo Scientific™ MicroCL 21 Microcentrifuge
GREENER_CHOICE
Save time with the fast sample processing of this microcentrifuge that provides productivity, versatility, safety and convenience in a compact, easy-to-use lab instrument. They are designed to provide acceleration up to 21,100xg in as little as 12 seconds.
Brand:  Thermo Scientific™ 75002549
2072.20 GBP valid until 2024-12-31
Use promo code "21629" to get your promotional price.
 View more versions of this product
Product Code. 17845537
  • £2619.06 / Each
Explore more special offers
Description
Description
Save time with the fast sample processing of Thermo Scientific MicroCL 21 microcentrifuges, providing productivity, versatility, safety and convenience in a compact, easy-to-use lab instrument. Lightweight microcentrifuge rotors are designed to provide acceleration up to 21,100xg in as little as 12 seconds.
Features:
  • Exceptional acceleration of up to 21,000xg in as little as 12 seconds
  • Choose from 7 optional microcentrifuge rotors to find the ideal fit for your application
    • Outstanding corrosion resistance with lightweight engineered polymer rotors
    • Standard rotor runs 24 microcentrifuge tubes in a single row, from 1.5 to 2.0 mL tubes to mini-preps and spin columns
    • Ergonomic one-click rotor opening and closing, as well as transparent design for added safety and convenience with Thermo Scientific ClickSeal biocontainment lids
    • Eliminate the need for adapters with unique dual row rotor
  • Simplify operation with intuitive controls, easy-to-read displays and fast one-click centrifuge lid closure
  • Sleek, lightweight, space-saving design
  • Durable and easy to clean and maintain
  • Quiet performance promotes a safe, stress-free environment
  • Conforms to the latest clinical and safety standards, including 98/79/EC In Vitro Diagnostic Directive
TRUSTED_SUSTAINABILITY
Specifications
Specifications
24 x 1.5/2 mL
21,100 x g
Ambient temperature
Benchtop Centrifuge
MicroCL 21 Centrifuge, Ventilated
2 Years
1 to 99 min. (1 min. increments); HOLD
8.9 x 9.6 x 13.8 in. (22.5 x 24.3 x 35.2 cm)
Thermo Scientific
230 V
14,800 rpm
56 dB(A)
CSA, CE, IVD
Dual Row 18 x 2.0/0.5mL rotor with screw-on lid
230 V 50/60 Hz
No
IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
23.1lb. (10.5 kg)
50/60 Hz
Dual Row 18 x 2/0.5 mL Rotor with Screw-On Lid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.