Learn More
Vileda Professional™ MicroControl Controlled Environments Mop
The MicroControl Mop is a reusable concept that offers improved quality and efficiency by reducing chemical costs in Controlled Environments up to ISO 5 (GMP A/B) on floors, walls and ceilings.
Brand: Vileda Professional™ 140708
Additional Details : Weight : 3.80000kg
Description
- 100% microfiber mop surface offering high cleaning performance
- Patented edge cleaning lip design allows for effective cleaning of corners, skirt boards etc.
- Straight and clean design with no dirt traps
- Multilayer design with sealed edges for high absorbency, low particle shedding and top stability
- Ergonomic performance and high efficiency due to the Vileda™ pre-prepared method
- Swap mopping system: One cleaning tool for floors, walls and ceilings
- Ideal microfiber cleaning results have been independently tested and validated
Specifications
Blue | |
Non-sterile | |
Reusable | |
Polyester |
Mop Pad | |
Autoclavable | |
No |