Learn More
Vileda Professional™ MicroEffective Controlled Environments Mop
MicroEffective CE Mop is a reusable concept with strong cleaning performance and low friction to clean less strict classification rooms. Compatible in ISO 8 to 9 areas with an improved version applicable for wider ISO applications (ISO 7 to 9).
Brand: Vileda Professional™ 161533
Additional Details : Weight : 3.60000kg
Description
- Efficient polyethersulfone-microfiber mop-cleaning performance coupled with extended coverage
- Hygienic, externally proven bacterial removal on floor, wall and ceiling surfaces
- Very low particle shedding of microfiber mops (Cat 1 for Hoods per IEST-RP-CC03.4); up to ISO 5 (GMP A/B)
- Enhanced decontamination result based on open side corners in the pockets
- Approved for CE usage/Designed for Sterilization (autoclaving/gamma irradiation)
- Ergonomic easy gliding mops and light weight mopping system
Specifications
Gray | |
Non-sterile | |
Reusable | |
Polyester |
Mop Pad | |
Autoclavable | |
No |