missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bolle Protection™ Microfiber Goggle Pouch
2.48 GBP valid until 2024-12-31
Use promo code "23287" to get your promotional price.
Use promo code "23287" to get your promotional price.
Click to view available options
Color:
Black
Specifications
Color | Black |
Product Type | Goggle Pouch |
For Use With (Application) | For Goggles |
Model | Etuifl |
No. per Pack | 1 Ea. |
Material | Microfiber |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'