missing translation for 'onlineSavingsMsg'
Learn More
Ansell MICROFLEX™ MIDKNIGHT™ XTRA 93-862 Nitrile Gloves with Extended Cuff Product Code.: 16409415
Add to basket

Ansell MICROFLEX™ MIDKNIGHT™ XTRA 93-862 Nitrile Gloves with Extended Cuff
GREENER_CHOICE

Product Code. 16409415
Medium
Click to view available options
Size:
2X-Large
Large
Medium
Small
X-Large

Product Code. 16409415

Brand: Ansell 93862080

Add to basket
Add to basket

Disposable nitrile gloves with extended cuff provide durable protection in the event of chemical splashes. NFPA and medical exam grade certified for use by healthcare professionals.

  • Tested against both fentanyl and gastric acid (vomit) to simulate authentic overdose situations
  • Dark, black color makes it easy to see when hands have come into contact with potentially hazardous powders
  • Made with a soft, flexible nitrile formulation
  • Fully textured for a reliable, consistent grip
  • For use with:
    • Administering drugs
    • Emergency services
    • Equipment repair and maintenance
    • Extra protection over wrist and forearm
    • Inspection, selecting, checking parts
    • Laboratory analysis and testing
    • Paint and spray shops
    • Patient examinations
    • Raw material sample collection
    • Tire rotation and change
Greener Choice Claims
  • Environmental benefits include:
    • Recyclable
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NFPA 1999:2018, ASTM D6319, FDA21 CFR 177-2600
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, EN 455 1-4
Applications Healthcare, Laboratory
Color Black
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 6.3 mil
Finger Thickness (Metric) 0.16 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.7 mil
Palm Thickness (Metric) 0.12 mm
Powdered Powder Free
Size Medium
Thickness (English) 6.3 mil
Thickness (Metric) 0.16 mm
Model 93862080
Series MICROFLEX™ MidKnight™ XTRA 93-862
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131

    Expires2025-06-08

    New Offer
    Shop Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell MICROFLEX™ MIDKNIGHT™ XTRA 93-862 Nitrile Gloves with Extended Cuff >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.