missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell Microflex™ NeoTouch™ 25-201 Disposable Neoprene Glove with Extended Cuff
Click to view available options
Size: X-Small
Description
- An easy-to-don Neoprene glove offering excellent chemical splash protection, and a reduced risk of Type 1 allergic reactions
- Designed with an extended cuff, it offers expanded protection over the wrist and forearm
- Textured fingertips help improve grip in wet and dry conditions
Specifications
Certifications/Compliance (Europe) | ASTM D6977|Category III|EN 421:2010|EN ISO 21420:2020|EN ISO 374-1:2016|EN ISO 374-5:2016|Please consult the IFU|ISO 13485|FDA21 CFR 177-2600-US Food Contact Approved |
Chlorinated | No |
Applications | Industrial, Laboratory |
Color | Green |
Latex Free | Yes |
Material | Neoprene |
Size | X-Small |
Thickness (English) | 7.9 mil |
Thickness (Metric) | 0.2 mm |
EN 421 | Yes |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'