missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes
Dispense Cassettes for use with MultiFlo FX Dispenser and EL406 Washer Dispenser.
Brand: Agilent BioTek 7170010
This item is not returnable.
View return policy
Description
- Volume range 1 μL to 10 mL
- Accommodates 6- to 1536-well microplates, PCR trays, deep-well microplates, microtubes and other tube configurations up to 4 in. in height
- Dispense 1536 wells in 18 s.
- Cassettes can be autoclaved without recalibration
- Dispense up to eight reagents at one time
- Customizable dispense pattern allows skipped columns or rows
- Built-in priming trough
- Easy maintenance of individually replaceable tips and choice of replacement cassettes or tubing
- Compatible with Bio-Stack microplate stacker 11-120-601 for increased throughput
Specifications
10 to 3000 μL | |
0.035 cm | |
10 μL | |
Large Volume Cassette |
0.014 in. | |
For liquid handling | |
Molded Polypropylene | |
EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes > Molded plastic tips; 10μL
Spot an opportunity for improvement?Share a Content Correction