missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek MultiFlo FX/EL406 Dispenser Cassettes
Shop All Agilent BioTek Products

Click to view available options
Diameter (Metric):
0.019 cm
0.02 cm
0.035 cm
Type:
Large Volume Cassette
Large Volume Replacement Tubing Set
Low Volume 1536 Well Cassette
Low Volume Cassette
Low Volume Replacement Tubing Set
Medium Volume Cassette
Medium Volume Replacement Tubing Set
Material:
Molded Polypropylene
Stainless Steel
Capacity (Metric):
1 to 50 μL
10 to 3000 μL
5 to 2500 μL
Unit Size:
Each
Description
- Volume range 1 μL to 10 mL
- Accommodates 6- to 1536-well microplates, PCR trays, deep-well microplates, microtubes and other tube configurations up to 4 in. in height
- Dispense 1536 wells in 18 s.
- Cassettes can be autoclaved without recalibration
- Dispense up to eight reagents at one time
- Customizable dispense pattern allows skipped columns or rows
- Built-in priming trough
- Easy maintenance of individually replaceable tips and choice of replacement cassettes or tubing
- Compatible with Bio-Stack microplate stacker 11-120-601 for increased throughput
Specifications
Specifications
| Capacity (Metric) | 5 to 2500 μL |
| Diameter (English) | 0.014 in. |
| Diameter (Metric) | 0.035 cm |
| For Use With (Application) | For liquid handling |
| Increments | 5 μL |
| Material | Molded Polypropylene |
| Type | Medium Volume Replacement Tubing Set |
| For Use With (Equipment) | EL406 Microplate Washer, Multiflo Dual Syringe Dispenser Module, Agilent BioTek MultiFlo FX Multimode Dispenser |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction