missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) MicroFunnel™ ST Filters Funnel with Supor Membrane

Microfunnel ST filter funnels are available in gamma-irradiated, double-bagged packaging saving valuable time during entry into cleanrooms and hoods.

Brand:  Cytiva (formerly Pall Lab) 4851

Additional Details : Weight : 0.00100kg

 View more versions of this product

Product Code. 17174421

  • £385.00 / Pack of 2
Estimated Shipment: 07-08-2024
to see stock.
Add to basket

Description

Description

  • For research use only
Specifications

Specifications

25 mm
0.2 μm
3.9 cm2
Gridded Filtering Funnel
100 mL
40/Pk.
Sterile
Supor (PES)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers