Learn More
Ansell Microgard™ 1500 Navy Coveralls with Hood
Product Code. 15536474 Shop All Ansell Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 40
Pack of 50
Product Code. 15536474
Brand: Ansell NV15S0013807
Only null left
Add to basket
Edge
Add to basket
Strip, clear up and handle asbestos safely. Ansell Microgard™ 1500 Coveralls with Hood filter particles as small as 3μ
  • Coverall with 3-piece hood
  • Elasticated wrists, waist, and ankles
  • 2-way front zipper with re-sealable storm flap
  • Filter 100% of particles >3 microns
  • Air and water vapor permeable to help reduce the risk of heat stress
  • Silicone-free for spray painting applications
  • Optimised body fit for wearer comfort and safety
TRUSTED_SUSTAINABILITY
Specifications
Product Type General Purpose Coveralls
Size 3X-Large
Gender Unisex
Material SMS Polypropylene
Color Navy
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel
Closure Type Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance (Europe) EN ISO 13982-1, EN13034
Model NV15-S-00-138
No. of Pockets None
Style Includes Hood