missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Microgard™ 1800 Standard Coveralls with Hood
214.04 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Coverall with 3-piece hood
- Elasticated wrists, waist and ankles
- 2-way front zipper with re-sealable storm flap
- Bound seams
- Standard: CE category III, type 5/6 EN 1149-5 (antistatic)
- Low hazard liquid spray, splash and particle protection
- Moisture vapour permeable (‘breathable’) to help reduce the risk of heat stress
- Silicone-free which is critical in spray painting applications
- Anti-static according to EN1149-5
- Low linting to reduce the risk of fibre contamination in some critical areas
- Optimised body fit for wearer comfort and safety
Specifications
Product Type | Protective Coveralls |
Size | Small |
Material | Microporous Polyethylene Film |
Color | White |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | 3-panel |
Closure Type | 2-way Front Zipper with Storm Flap |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'