Learn More
Ansell Microgard™ 2000 Standard Overshoes
364.91 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Description
- Microporous polyethylene laminate
- Elasticated opening
- Bound seams
- Barrier protection from hazardous liquid and particulates
Specifications
Material | Polyethylene |
Color | White |
Closure Type | Elastic Opening |
Model | WH20-B-00-400-00 |
Seam Type | Bound |
Packaging Quantity | 400/Pk |
Product Type | Shoe Covers |