missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ Microgard™ 2000 Ts Plus Coveralls with Hood
369.47 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Proven barrier to low concentration liquid chemicals, diluted pesticides, liquid and particulate biological hazards
- Moisture vapour permeable (‘breathable’) to help reduce the risk of heat stress
- Silicone-free for spray painting applications
- Low Linting to reduce the risk of contamination in critical areas
- Anti-static according to EN 1149-1
- Optimized body fit for wearer comfort and safety
- Tunnelled elasticated wrists, hood and ankles minimises the risk of linting and cross contamination
- Thumb loops prevent sleeve movement when working above overhead
- Chinstrap helps to reduce the risk of cross contamination
Specifications
Product Type | Protective Coveralls |
Size | 3X-Large |
Material | Microporous Polyethylene Film |
Color | White |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | 3-panel, Elastic |
Closure Type | Zipper with Re-sealable Flap |
Seam Type | Stitched, Taped |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'