Learn More
Ansell Microgard™ 2500 Standard SOCO Overboots
457.89 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Description
- Microporous polypropylene laminate
- Elastic to top of boot
- Tie fastening
- Blue binding to seams
- Reinforced Surestep non-slip soles
- Adjustable shoe tie
- Fits Euro sizes 42 to 46 (UK 8 to 11)
Specifications
Material | Polypropylene |
Color | White |
Closure Type | Elastic Top |
Sole Type | Non-slip |
Model | WH25-B-00-409-00 |
Seam Type | Bound with Blue binding to seams |
Packaging Quantity | 100 Pairs |
Product Type | Boot Covers |