Learn More
Ansell Microgard™ 3000 Protective Overshoes
627.19 GBP valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Description
- Multi-layer barrier laminate
- Elasticated opening
- Fits over Euro sizes 42 to 46. (UK 8 to 11)
Specifications
Size | Fits over Euro shoe sizes 42 to 46 (UK 8 to 11) |
Material | Multi-layer Barrier Laminate Fabric |
Color | Yellow |
Closure Type | Elastic Top |
Seam Type | Ultrasonically Welded |
Certifications/Compliance (Europe) | Category I PPE according to EuropeanDirective 89/686/EEC |
Packaging Quantity | 250 Pairs |
Product Type | Shoe Covers |