Learn More
Ansell™ Microgard™ FR Coveralls with Hood
Product Code. 15576424 Shop All Ansell Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Pack of 30
Microgard™ FR should never be worn in isolation for flame retardant protection. Always wear over the top of garments which achieve EN533 Index 2 or above.
Product Code. 15576424
Brand: Ansell™ BL95S0011105
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Microgard™ FR should never be worn in isolation for flame retardant protection. Always wear over the top of garments which achieve EN533 Index 2 or above.
Designed to be worn over woven thermal protective garments such as NOMEX™ Ansell Microgard™ FR is a flame retardant material offering protection from particulates and light liquid splash without compromising worker protection in the event of a flash fire*.
  • Coverall with 2-piece hood, stitched seams, elasticated hood, wrists, waist and ankles
  • 2-way front zipper with storm flap
  • Flame retardant treated Sontara/wood pulp/polyester fabric
  • 3-thread overlocked seams
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Flame-resistant Coveralls
Size X-Large
Gender Unisex
Material Multi-layer Barrier Laminate Fabric
Color Blue
Disposable Limited-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 2-panel
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance Cat. III Type 5 & 6, EN533 Index 1, EN1073-2, EN1149-1
No. of Pockets None
Style Includes Hood