missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novacyt Group Microgen Bioproducts™ SARS-CoV-2 IgG EIA
For the screening of patients previously infected, to either; confirm infection, monitor the course of disease, or to draw epidemiological conclusions. Includes all additional reagents required to perform each test.
Brand: Novacyt Group MBCOV2CE
This item is not returnable.
View return policy
Description
- The Microgen Bioproducts™ SARS-CoV-2 IgG EIA introduces a highly sensitive and specific assay for the qualitative detection of IgG antibodies based on the nucleocapsid protein, usually detected two weeks after initial infection
- The test is intended for the screening of patients previously infected, to either; confirm infection, monitor the course of disease, or to draw epidemiological conclusions
- Each kit contains 480 reactions (5 x 96 well plates) and includes all additional reagents required to perform each test –alongside positive/negative controls. Sample types include human serum and plasma
- Convenient 480 reaction format (5 x 96 well plates) including all plates and additional reagents to perform the assay
- Compliant with The Medicines and Healthcare Product Regulatory Agency Ministry (MHRA) TPP
- High Sensitivity and Specificity
- Less than 1 hour 30 minutes incubation time
- Functional with human plasma and serum samples
- No cross-reactivity with common viral pathogens
- Validated for manual application, however also suitable for high-throughput ELISA automation
- Vital in the process of monitoring COVID-19 antibodies
Specifications
Standard ELISA calibration format - Pos/neg controls included with the kit | |
2°C to 8°C | |
Yes | |
Enzyme-linked immunosorbent assay; chromatography/colour change | |
Human Plasma or Human Serum | |
Antibody ELISA | |
ELISA | |
Positive control 1 x 3.5 mL, Negative control 1 x 3.5 mL, Wash buffer 2 x 12 5 mL, Sample buffer 2 x 125 mL, Conjugate 1 x 60 mL, Substrate solution 1 x 60 mL, Stop solution 1 x 60 mL | |
480 | |
Foil pouches and plastic bottles | |
5 μL of human plasma or human serum | |
0.994 | |
Antibodies/Nucleocapsid protein |
CE IVD certified, DoC Documentation | |
Both positive and negative controls | |
SARS-CoV-2 IgG Antibodies (Nucleocapsid Protein) | |
96-well Plate | |
100% - > 14 days post PCR confirmation | |
Chemical compositions are found in Safety Data Sheets | |
Manual OR Automated ELISA Systems & Plate Reader for Interpretation | |
Polypropylene Plates | |
Antibodies | |
15/Pk. | |
Human | |
Sterile | |
<1.5 hr. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novacyt Group Microgen Bioproducts™ SARS-CoV-2 IgG EIA > 480 Reactions
Spot an opportunity for improvement?Share a Content Correction