Learn More
Fisherbrand™ Microglass Fiber Filter Discs
Made with 100% borosilicate micro-glass fibers. Fisherbrand™ Microglass Fiber Filter Discs are non-hygroscopic and biologically inert. They show chemical resistance to most solvents and reagents (except hydrofluoric acid).
Brand: Fisherbrand™
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Contains no binder
- Suitable for use at and up to 500°C
- Not affected by humidity
- Suitable for micro-filtration due to fine fibers
- High-loading capacity
- High chemical purity
- made with demineralized water
- High permeability to air
- Filtration of industrial water, fresh water algae, drinking water, Proteins, samples for HPLC, solvents
- Sediments content in petroleum products
- Ligand binding test
- Air pollution monitoring
- Membranes pre-filters
- Radio-immuno tests
- Suspended solids in water, smokes
- Liquid scintillation counting
- Cells collection
- Grease content in dairy products
Specifications
75 g/m2 | |
Disc | |
0.7 μm | |
310 sec. | |
263 | |
Non-hygroscopic | |
0.45 mm | |
37 mm |
Microfilter | |
Borosilicate Glass Microfiber | |
0.7 μm | |
Staining of dyed Papers, purification filtration, solvents filtration, samples filtration HPLC, Proteins filtration, cells collection, critical analysis | |
100 Pack | |
500°C | |
Filter |