missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microglass Fiber Filters, Grade 227
Suitable for micro-filtration and can be used in various fields.
£5.50 - £15.60
Specifications
Basis Weight | 70 g/m2 |
---|---|
Filter Type | Microfilter |
Format | Disc |
Material | Borosilicate Glass |
Pore Size | 0.7 to 2.7 μm |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
11744586
|
Fisherbrand™
|
25 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11774586
|
Fisherbrand™
|
70 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11784586
|
Fisherbrand™
|
90 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- High level of particle retension
- Large load capacity
- High chemical purity (made with demineralized water)
- Non-hygroscopic and biologically inert
- High permeability to air
- Chemical resistance to most solvents and reagents (except hydrofluoric acid)
- Resist to temperature of about 500°C
Specifications
70 g/m2 | |
Disc | |
0.7 to 2.7 μm | |
227 | |
500°C | |
0.4 mm |
Microfilter | |
Borosilicate Glass | |
Air pollution control, separation of soot before gas analyzers, support weighing in the balance analyzes and dryers | |
Non-hygroscopic | |
0.015 in. | |
Filter |