missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Microglass Fiber Filters, Grade 259

Made with 100% borosilicate micro-glass fibers. Fisherbrand™ Microglass Fiber Filters, Grade 259 are non-hygroscopic and biologically inert. They show chemical resistance to most solvents and reagents (except hydrofluoric acid).

Brand:  Fisherbrand™

Code : 47

Additional Details : Weight : 0.10080kg

 View more versions of this product

Product Code. 11475238

  • £104.00 / Pack of 100
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • Contains no binder
  • Suitable for use at and up to 500°C
  • Not affected by humidity
  • Suitable for micro-filtration due to fine fibers
  • High-loading capacity
  • High chemical purity
  • made with demineralized water
  • High permeability to air
Applications:
  • Filtration of industrial water, fresh water algae, drinking water, Proteins, samples for HPLC, solvents
  • Sediments content in petroleum products
  • Ligand binding test
  • Air pollution monitoring
  • Membranes pre-filters
  • Radio-immuno tests
  • Suspended solids in water, smokes
  • Liquid scintillation counting
  • Cells collection
  • Grease content in dairy products
Specifications

Specifications

52 g/m2
Disc
1.6 μm
Fresh water algae, sediments content (petroleum products), solvents filtration, liquid scintillation counting, suspended solids in smokes, grease content in dairy products
Non-hygroscopic
0.01 in.
Filter
Microfilter
Borosilicate Glass
60 sec.
259
500°C
0.26 mm
109 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers