Learn More
Fisherbrand™ Microglass Fiber Filters, Grade 259
Made with 100% borosilicate micro-glass fibers. Fisherbrand™ Microglass Fiber Filters, Grade 259 are non-hygroscopic and biologically inert. They show chemical resistance to most solvents and reagents (except hydrofluoric acid).
Brand: Fisherbrand™
Code : 47
Additional Details : Weight : 0.03008kg
Description
- Contains no binder
- Suitable for use at and up to 500°C
- Not affected by humidity
- Suitable for micro-filtration due to fine fibers
- High-loading capacity
- High chemical purity
- made with demineralized water
- High permeability to air
- Filtration of industrial water, fresh water algae, drinking water, Proteins, samples for HPLC, solvents
- Sediments content in petroleum products
- Ligand binding test
- Air pollution monitoring
- Membranes pre-filters
- Radio-immuno tests
- Suspended solids in water, smokes
- Liquid scintillation counting
- Cells collection
- Grease content in dairy products
Specifications
52 g/m2 | |
Disc | |
1.6 μm | |
Fresh water algae, sediments content (petroleum products), solvents filtration, liquid scintillation counting, suspended solids in smokes, grease content in dairy products | |
Non-hygroscopic | |
0.01 in. | |
Filter |
Microfilter | |
Borosilicate Glass | |
60 sec. | |
259 | |
500°C | |
0.26 mm | |
38 mm |