Learn More
Hamilton™ Microlab™ 600 Diluter/Dispenser System Upgrade Kit
Unlock Advanced functionality of your Microlab 600. Simply plug in this easy-to-use Hamilton SD card to instantly transform your Basic Microlab 600 into an Advanced version.
Brand: Hamilton™ 61500-02
Code : NEW
Additional Details : Weight : 0.35200kg
Specifications
Microlab 600 | |
Basic to Advanced Controller Upgrade Kit | |
Advanced Manual, 2 GB SD Card, SD to USB Converter, Programmer Software CD |