Learn More
Hamilton™ Microliter™ and Gastight™ Syringes: Model 701 for Agilent 7673, 7683, 7693 and 6850 ALS GC
Designed to work with Agilent 7673, 7683, 7693 and 6850 Automatic Liquid Sampler (ALS) gas chromatography (GC) autosamplers
£57.65 - £767.00
Specifications
Material Borosilicate Glass (Barrel), Stainless Steel (Plunger)
Diameter (English) Outer 0.26 in.
Diameter (Metric) Outer 6.6 mm
Length (English) Needle 1.71 in.
Product Type Microliter Syringe
View More Specs
Products 11
Product Code Brand Needle Gauge Model Needle Point Number Quantity Price Quantity & Availability  
Product Code Brand Needle Gauge Model Needle Point Number Quantity Price Quantity & Availability  
10722711
View Documents
Hamilton™
80390
23s G 701 N (Cemented Needle) AS 6 Pk.
£591.00
Pack of 6
10508502
View Documents
Hamilton™
80357
23s G 701 RN (Removable Needle) AS Ea.
£99.55
Each
10340281
View Documents
Hamilton™
80393
23 to 26s G 701 N (Cemented Needle) AS Ea.
£80.65
Each
11567614
View Documents
Hamilton™
80399
26s G 701 N (Cemented Needle) 2 Ea.
£60.30
Each
10376741
missing translation for 'viewDocuments'
Hamilton™
80387
23s G 701 N (Cemented Needle) AS Ea.
£65.20
Each
10300331
missing translation for 'viewDocuments'
Hamilton™
80359
23 to 26s G 701 RN (Removable Needle) AS Ea.
£96.75
Each
10117070
missing translation for 'viewDocuments'
Hamilton™
80391
23 to 26s G 701 N (Cemented Needle) AS 6 Pk.
£767.00
Pack of 6
10156970
missing translation for 'viewDocuments'
Hamilton™
80388
26s G 701 N (Cemented Needle) AS Ea.
£57.65
Each
10442721
missing translation for 'viewDocuments'
Hamilton™
80358
26s G 701 RN (Removable Needle) AS Ea.
£104.00
Each
10347012
missing translation for 'viewDocuments'
Hamilton™
80389
26s G 701 N (Cemented Needle) AS 6 Pk.
£345.00
Pack of 6
10055630
missing translation for 'viewDocuments'
Hamilton™
80398
23s G 701 N (Cemented Needle) 2 Ea.
£65.50
Each
Beschreibung
Beschreibung
Microliter syringes have a stainless steel plunger that is hand fitted to the glass barrel and are designed for liquid sampling.
Syringe Terminations
  • Cemented Needle (N) - For low volume syringes the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
  • Removable Needle (RN) - The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Needle Point Styles
  • Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 23s or 26s
  • Point Style AS - Special conical style needle point used on autosampler syringes the non-coring needle point is recommended for septum penetration. Available gauges: 23s, 26s, 23s-26s
Spezifikation
Spezifikation
Borosilicate Glass (Barrel), Stainless Steel (Plunger)
6.6 mm
Microliter Syringe
10°C
Agilent 7673, 7683, 6850 Automatic Liquid Sampler, Gas Chromatography Autosamplers
0.26 in.
1.71 in.
50°F
10 μL
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt