missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One MICROLON™ 600 V-Bottom 96-well ELISA Microplate
GREENER_CHOICE

Product Code. 10560783
Click to view available options
Color:
Clear
Unit Size:
Pack of 40
This item is not returnable. View return policy

Product Code. 10560783

Brand: Greiner Bio-One 651061

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Free of detectable DNase, RNase, human DNA and pyrogens

Alphanumeric well coding
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens
Format Standard
Lid Without Lid
Array 8 x 12
Autoclavable Not Autoclavable
Binding Type High Binding
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 40 to 200 μL
Sterility Non-sterile
Well Shape Conical Bottom (V-well)
Dnase Free Yes
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 40
No. per Pack 10
RNase Free Yes
Dimensions (L x W x H) 128 x 85 x 14 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.