Learn More
Greiner Bio-One Microlon™ Polystyrene 96-well ELISA Microplates
GREENER_CHOICE
Product Code. 10043211
Click to view available options
Binding Property:
High Binding
Medium Binding
Surface Treatment:
Microlon 200
Microlon 600
Well Design:
Chimney Well
Standard Well
Unit Size:
Pack of 40
Product Code. 10043211
Brand: Greiner Bio-One 655081
Only null left
Add to basket
Add to basket
Manufactured from crystal clear polystyrene for optimum visibility. Greiner BioOne™ Microlon™ Polystyrene 96-Well ELISA Microplates are tested in a colorimetric immunoasay for immunological research applications.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Tracebility from production through a defined LOT number system
  • Manufactured from crystal clear polystyrene
  • The footprint is compatible with automated systems
  • Well to well spacing : 9 mm
  • Alphanumeric well coding
  • Suitable for transmission measurements
  • Specifically treated to provide an incresed protein binding
  • Well profile: F-bottom / standard (flat bottom) 
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Medium Binding
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Microlon 600
Well Shape Flat Bottom (F-well)
Quantity 40/Cs.
Well Design Chimney Well