missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Microlon™ Polystyrene 96-well ELISA Microplates
Manufactured from crystal clear polystyrene for optimum visibility. Greiner BioOne™ Microlon™ Polystyrene 96-Well ELISA Microplates are tested in a colorimetric immunoasay for immunological research applications.
Brand: Greiner Bio-One 655080
This item is not returnable.
View return policy
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Tracebility from production through a defined LOT number system
- Manufactured from crystal clear polystyrene
- The footprint is compatible with automated systems
- Well to well spacing : 9 mm
- Alphanumeric well coding
- Suitable for transmission measurements
- Specifically treated to provide an incresed protein binding
- Well profile: F-bottom / standard (flat bottom)
Specifications
Medium Binding | |
Without Lid | |
Clear | |
96 | |
Microlon 200 | |
40/Cs. |
Standard | |
8 x 12 | |
Polystyrene | |
Non-sterile | |
Flat Bottom (F-well) | |
Chimney Well |