Learn More
Epredia™ Microm™ Cryostats Heat Extractor
Use Epredia™ Microm™ Cryostats Heat Extractors with Epredia™ Cryostats.
£109.00 - £252.00
Specifications
Product Type Heat Extractor
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
12033059
View Documents Promotion Details
Epredia™
524510
HM 550, HM 560 Series
£109.00
Each
12023059
View Documents
Epredia™
524490
HM 525
£252.00
Each
Description
Description
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen
  • Can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on an active Peltier
Specifications
Specifications
Heat Extractor